Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017067.1 | NP_058763.1 | 4 - 5 | 313 | 145 | |
GenBank mRNA | L13722.1 | - | 4 - 5 | 313 | 145 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |