Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108244.1 | NP_001101714.1 | 1 - 2 | 419 | 100 | |
XM_006257199.2 | XP_006257261.1 | 1 - 2 | 848 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |