Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ell2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008760603.1 | XP_008758825.1 | 9 - 10 | 2311 | 135 | |
XM_008760604.1 | XP_008758826.1 | 9 - 10 | 1597 | 135 | ||
GenBank mRNA | FQ230057.1 | - | 2 - 3 | 305 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |