Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013984.1 | NP_001014006.1 | 4 - 5 | 304 | 65 | |
XM_006249997.2 | XP_006250059.1 | 4 - 5 | 452 | 65 | ||
GenBank mRNA | BC082004.1 | - | 4 - 5 | 304 | 7131519 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |