Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fmo3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053433.2 | NP_445885.2 | 2 - 3 | 357 | 174 | |
GenBank mRNA | AF286595.1 | - | 2 - 3 | 201 | 174 | |
BC087008.1 | - | 2 - 3 | 357 | 7129948 | 174 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons |