Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113789.1 | NP_001107261.1 | 3 - 4 | 649 | 135 | |
NM_001143756.1 | NP_001137228.1 | 3 - 4 | 488 | 135 | ||
XM_006230495.2 | XP_006230557.1 | 3 - 4 | 562 | 135 | ||
XM_008760008.1 | XP_008758230.1 | 3 - 4 | 1390 | 135 | ||
XM_008760009.1 | XP_008758231.1 | 3 - 4 | 529 | 135 | ||
GenBank mRNA | BC158629.1 | - | 2 - 3 | 386 | 7456037 | 135 |
CK469366.1 | - | 3 - 4 | 488 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |