Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ighmbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031586.1 | NP_113774.1 | 5 - 6 | 709 | 83 | |
GenBank mRNA | AF199411.1 | - | 5 - 6 | 709 | 83 | |
BC099790.1 | - | 5 - 6 | 905 | 7934952 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |