Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008759409.1 | XP_008757631.1 | 2 - 3 | 923 | 72 | |
XM_008759410.1 | XP_008757632.1 | 2 - 3 | 2000 | 72 | ||
XM_008759411.1 | XP_008757633.1 | 2 - 3 | 870 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |