Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prr12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008759481.1 | XP_008757703.1 | 8 - 9 | 5120 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |