Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107501.1 | NP_001100971.1 | 8 - 9 | 1248 | 98 | |
XM_008759148.1 | XP_008757370.1 | 8 - 9 | 1279 | 98 | ||
XM_008759149.1 | XP_008757371.1 | 8 - 9 | 1264 | 98 | ||
XM_008759150.1 | XP_008757372.1 | 8 - 9 | 1267 | 98 | ||
XM_008759151.1 | XP_008757373.1 | 8 - 9 | 1252 | 98 |
Gene Symbol: LOC100911911 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003748820.3 | XP_003748868.1 | 8 - 9 | 1279 | 98 | |
XM_008759239.1 | XP_008757461.1 | 8 - 9 | 1280 | 98 | ||
XM_008759240.1 | XP_008757462.1 | 8 - 9 | 1264 | 98 | ||
XM_008759241.1 | XP_008757463.1 | 8 - 9 | 1268 | 98 | ||
XM_008759242.1 | XP_008757464.1 | 8 - 9 | 1252 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |