Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capns1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017118.1 | NP_058814.1 | 663 | 91 | ||
GenBank mRNA | BC098068.1 | - | 663 | 7462537 | 91 | |
U10861.1 | - | 5 - 6 | 352 | 91 | ||
U53859.1 | - | 599 | 91 |
Gene Symbol: LOC100911363 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003748836.3 | XP_003748884.1 | 8 - 9 | 712 | 91 | |
XM_008759263.1 | XP_008757485.1 | 8 - 9 | 914 | 91 |
Gene Symbol: LOC100912380 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006228796.2 | XP_006228858.2 | 7 - 8 | 439 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |