Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017359.2 | NP_059055.2 | 1 - 2 | 174 | 94 | |
GenBank mRNA | AY724493.1 | - | 1 - 2 | 353 | 94 | |
BC085744.1 | - | 1 - 2 | 174 | 7191921 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |