Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Colec11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006239960.2 | XP_006240022.1 | 2 - 3 | 388 | 151 | |
XM_006239961.1 | XP_006240023.1 | 2 - 3 | 181 | 151 | ||
XM_006239962.2 | XP_006240024.1 | 2 - 3 | 387 | 151 | ||
GenBank mRNA | BC158664.1 | - | 184 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |