Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hpcal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017356.2 | NP_059052.1 | 2 - 3 | 311 | 102 | |
XM_006239914.2 | XP_006239976.1 | 3 - 4 | 918 | 102 | ||
XM_006239916.2 | XP_006239978.1 | 2 - 3 | 813 | 102 | ||
XM_008764605.1 | XP_008762827.1 | 2 - 3 | 428 | 102 | ||
XM_008764606.1 | XP_008762828.1 | 4 - 5 | 1028 | 102 | ||
GenBank mRNA | BC088759.1 | - | 2 - 3 | 311 | 7317905 | 102 |
D13126.1 | - | 2 - 3 | 266 | 102 | ||
FQ220970.1 | - | 2 - 3 | 301 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |