Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sntg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106720.1 | NP_001100190.1 | 9 - 10 | 955 | 77 | |
XM_006239967.2 | XP_006240029.1 | 13 - 14 | 1236 | 77 | ||
XM_006239968.2 | XP_006240030.1 | 12 - 13 | 1116 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |