Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105925.1 | NP_001099395.1 | 5 - 6 | 1004 | 134 | |
XM_008769195.1 | XP_008767417.1 | 5 - 6 | 2287 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Protease Rattus norvegicus Probe spans exons |