Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cryba4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031689.1 | NP_113877.1 | 3 - 4 | 194 | 79 | |
GenBank mRNA | AF013247.1 | - | 3 - 4 | 194 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |