Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Braf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006236357.2 | XP_006236419.2 | 13 - 14 | 1621 | 80 | |
XM_006236358.2 | XP_006236420.2 | 13 - 14 | 1537 | 80 | ||
XM_006236359.2 | XP_006236421.2 | 12 - 13 | 1501 | 80 | ||
XM_231692.8 | XP_231692.4 | 15 - 16 | 1663 | 80 | ||
GenBank mRNA | AF352172.1 | - | 1 - 2 | 64 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Apoptosis Protein Kinase Rattus norvegicus Inventoried Probe spans exons |