Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Braf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006236357.2 | XP_006236419.2 | 16 - 17 | 1968 | 104 | |
XM_006236358.2 | XP_006236420.2 | 16 - 17 | 1884 | 104 | ||
XM_006236359.2 | XP_006236421.2 | 15 - 16 | 1848 | 104 | ||
XM_231692.8 | XP_231692.4 | 18 - 19 | 2010 | 104 | ||
GenBank mRNA | AF352172.1 | - | 4 - 5 | 411 | 104 | |
FQ232056.1 | - | 3 - 4 | 334 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Apoptosis Protein Kinase Rattus norvegicus 1700 Inventoried Probe spans exons |