Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Braf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006236357.2 | XP_006236419.2 | 1 - 2 | 117 | 132 | |
XM_006236358.2 | XP_006236420.2 | 1 - 2 | 117 | 132 | ||
XM_006236359.2 | XP_006236421.2 | 1 - 2 | 117 | 132 | ||
XM_231692.8 | XP_231692.4 | 1 - 2 | 222 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Apoptosis Protein Kinase Rattus norvegicus Probe spans exons |