Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013083.2 | NP_001013101.2 | 7 - 8 | 691 | 108 | |
GenBank mRNA | CF249031.1 | - | 4 - 5 | 377 | 108 | |
CF249511.1 | - | 1 - 2 | 55 | 108 | ||
FQ226028.1 | - | 4 - 5 | 374 | 108 | ||
FQ234275.1 | - | 3 - 4 | 240 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Rattus norvegicus Probe spans exons |