Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013083.2 | NP_001013101.2 | 8 - 9 | 784 | 74 | |
GenBank mRNA | CF249031.1 | - | 5 - 6 | 470 | 74 | |
CF249511.1 | - | 2 - 3 | 148 | 74 | ||
FQ226028.1 | - | 5 - 6 | 467 | 74 | ||
FQ234275.1 | - | 4 - 5 | 333 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Rattus norvegicus Probe spans exons |