Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012985.2 | NP_037117.1 | 2 - 3 | 213 | 86 | |
GenBank mRNA | BC059148.1 | - | 2 - 3 | 76 | 6919202 | 86 |
D86215.1 | - | 2 - 3 | 213 | 86 | ||
FQ228773.1 | - | 2 - 3 | 74 | 86 | ||
FQ229037.1 | - | 2 - 3 | 73 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |