Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pon3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004086.1 | NP_001004086.1 | 5 - 6 | 508 | 117 | |
GenBank mRNA | BC079466.1 | - | 5 - 6 | 508 | 7122531 | 117 |
FQ218322.1 | - | 2 - 3 | 247 | 117 | ||
FQ218352.1 | - | 2 - 3 | 247 | 117 | ||
FQ218880.1 | - | 2 - 3 | 247 | 117 | ||
FQ218895.1 | - | 2 - 3 | 248 | 117 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |