Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wnt16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109223.1 | NP_001102693.1 | 1 - 2 | 380 | 60 | |
XM_006236143.2 | XP_006236205.1 | 1 - 2 | 429 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |