Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134994.1 | NP_001128466.1 | 2 - 3 | 208 | 144 | |
GenBank mRNA | BC168695.1 | - | 2 - 3 | 97 | 7938123 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |