Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Athl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006230587.2 | XP_006230649.1 | 9 - 10 | 1986 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |