Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nap1l4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012170.1 | NP_001012170.1 | 8 - 9 | 714 | 126 | |
XM_006230823.2 | XP_006230885.1 | 8 - 9 | 720 | 126 | ||
XM_006230824.2 | XP_006230886.1 | 9 - 10 | 787 | 126 | ||
XM_006230826.2 | XP_006230888.1 | 8 - 9 | 720 | 126 | ||
GenBank mRNA | BC085801.1 | - | 8 - 9 | 714 | 7110617 | 126 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |