Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_576263.6 | XP_576263.2 | 1 - 2 | 128 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Rattus norvegicus Probe spans exons |