Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gsdmd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130553.1 | NP_001124025.1 | 6 - 7 | 827 | 129 | |
XM_006241809.2 | XP_006241871.1 | 6 - 7 | 1482 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |