Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171981.1 | NP_001165452.1 | 10 - 11 | 926 | 69 | |
XM_008765682.1 | XP_008763904.1 | 9 - 10 | 1991 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |