Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpsf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106785.1 | NP_001100255.1 | 6 - 7 | 1238 | 108 | |
XM_006241372.1 | XP_006241434.1 | 7 - 8 | 1381 | 108 | ||
XM_006241373.1 | XP_006241435.1 | 7 - 8 | 1381 | 108 | ||
XM_006241374.1 | XP_006241436.1 | 7 - 8 | 1381 | 108 | ||
XM_008765386.1 | XP_008763608.1 | 7 - 8 | 1381 | 108 | ||
XM_008765387.1 | XP_008763609.1 | 6 - 7 | 1270 | 108 | ||
XM_008765388.1 | XP_008763610.1 | 6 - 7 | 1270 | 108 | ||
XM_008765390.1 | XP_008763612.1 | 7 - 8 | 1306 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |