Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gas2l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008765305.1 | XP_008763527.1 | 1 - 2 | 1975 | 63 | |
XM_008765306.1 | XP_008763528.1 | 1 - 2 | 1975 | 63 | ||
XM_008765307.1 | XP_008763529.1 | 1 - 2 | 1975 | 63 | ||
XM_008765308.1 | XP_008763530.1 | 1 - 2 | 1975 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Apoptosis Rattus norvegicus Probe spans exons |