Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133526.1 | NP_598210.1 | 4 - 5 | 568 | 76 | |
GenBank mRNA | FQ214867.1 | - | 4 - 5 | 476 | 76 | |
FQ216712.1 | - | 4 - 5 | 597 | 76 | ||
FQ227464.1 | - | 4 - 5 | 585 | 76 | ||
FQ229522.1 | - | 4 - 5 | 486 | 76 | ||
Y13275.2 | - | 4 - 5 | 568 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |