Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kitlg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021843.4 | NP_068615.1 | 3 - 4 | 365 | 115 | |
NM_021844.2 | NP_068616.1 | 3 - 4 | 365 | 115 | ||
XM_008765339.1 | XP_008763561.1 | 3 - 4 | 409 | 115 | ||
XM_008765340.1 | XP_008763562.1 | 3 - 4 | 409 | 115 | ||
GenBank mRNA | AB105879.1 | - | 3 - 4 | 365 | 115 | |
AF071204.1 | - | 3 - 4 | 200 | 115 | ||
AF071205.1 | - | 3 - 4 | 201 | 115 | ||
CB751462.1 | - | 3 - 4 | 365 | 115 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |