Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gas2l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001056437.5 | XP_001056437.2 | 6 - 7 | 2311 | 87 | |
XM_006241237.2 | XP_006241299.1 | 6 - 7 | 639 | 87 | ||
XM_008765305.1 | XP_008763527.1 | 6 - 7 | 3095 | 87 | ||
XM_008765306.1 | XP_008763528.1 | 7 - 8 | 2626 | 87 | ||
XM_008765307.1 | XP_008763529.1 | 7 - 8 | 3181 | 87 | ||
XM_008765308.1 | XP_008763530.1 | 7 - 8 | 2629 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Apoptosis 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |