Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnptab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_343194.7 | XP_343195.3 | 20 - 21 | 3833 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Rattus norvegicus 1700 Probe spans exons |