Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199098.1 | NP_954529.1 | 3 - 4 | 666 | 81 | |
XM_006241347.2 | XP_006241409.1 | 4 - 5 | 763 | 81 | ||
XM_006241348.2 | XP_006241410.1 | 4 - 5 | 616 | 81 | ||
XM_006241349.1 | XP_006241411.1 | 4 - 5 | 501 | 81 | ||
XM_008765355.1 | XP_008763577.1 | 3 - 4 | 712 | 81 | ||
GenBank mRNA | BC061871.1 | - | 3 - 4 | 666 | 5598899 | 81 |
Gene Symbol: LOC100912615 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008765371.1 | XP_008763593.1 | 771 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |