Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc6a15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172321.1 | NP_758824.1 | 5 - 6 | 1647 | 125 | |
GenBank mRNA | L22022.1 | - | 5 - 6 | 1647 | 125 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Rattus norvegicus Probe spans exons |