Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108094.2 | NP_001101564.1 | 8 - 9 | 1046 | 82 | |
GenBank mRNA | BC128768.1 | - | 8 - 9 | 1046 | 7125536 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |