Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: E2f7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108092.1 | NP_001101562.1 | 4 - 5 | 877 | 91 | |
XM_006241320.2 | XP_006241382.1 | 4 - 5 | 768 | 91 | ||
XM_006241321.2 | XP_006241383.1 | 4 - 5 | 610 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |