Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012619.2 | NP_036751.2 | 3 - 4 | 439 | 66 | |
GenBank mRNA | BC078881.1 | - | 3 - 4 | 439 | 7131414 | 66 |
FQ219680.1 | - | 3 - 4 | 469 | 66 | ||
M12337.1 | - | 3 - 4 | 424 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |