Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab3ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017313.3 | NP_059009.2 | 3 - 4 | 637 | 71 | |
XM_006241357.2 | XP_006241419.1 | 4 - 5 | 806 | 71 | ||
XM_006241358.2 | XP_006241420.1 | 4 - 5 | 811 | 71 | ||
XM_006241359.2 | XP_006241421.1 | 4 - 5 | 1815 | 71 | ||
XM_006241360.2 | XP_006241422.1 | 4 - 5 | 801 | 71 | ||
XM_008765383.1 | XP_008763605.1 | 3 - 4 | 246 | 71 | ||
GenBank mRNA | BC127500.1 | - | 3 - 4 | 640 | 7457420 | 71 |
U19181.1 | - | 3 - 4 | 630 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |