Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab3ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017313.3 | NP_059009.2 | 4 - 5 | 718 | 78 | |
XM_006241357.2 | XP_006241419.1 | 5 - 6 | 887 | 78 | ||
XM_006241358.2 | XP_006241420.1 | 5 - 6 | 892 | 78 | ||
XM_006241359.2 | XP_006241421.1 | 5 - 6 | 1896 | 78 | ||
XM_006241360.2 | XP_006241422.1 | 5 - 6 | 882 | 78 | ||
XM_008765382.1 | XP_008763604.1 | 3 - 4 | 848 | 78 | ||
XM_008765383.1 | XP_008763605.1 | 4 - 5 | 327 | 78 | ||
GenBank mRNA | BC127500.1 | - | 4 - 5 | 721 | 7457420 | 78 |
U19181.1 | - | 4 - 5 | 711 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |