Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gli1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191910.1 | NP_001178839.1 | 3 - 4 | 387 | 75 | |
XM_006241442.2 | XP_006241504.1 | 5 - 6 | 652 | 75 | ||
XM_006241443.2 | XP_006241505.1 | 4 - 5 | 535 | 75 | ||
XM_008765375.1 | XP_008763597.1 | 5 - 6 | 600 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |