Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chadl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008765807.1 | XP_008764029.1 | 3 - 4 | 2169 | 73 | |
XM_008765808.1 | XP_008764030.1 | 3 - 4 | 2169 | 73 | ||
XM_008765809.1 | XP_008764031.1 | 4 - 5 | 3509 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus Probe spans exons |