Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhpn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006241900.2 | XP_006241962.1 | 3 - 4 | 470 | 95 | |
XM_006241901.2 | XP_006241963.1 | 3 - 4 | 471 | 95 | ||
XM_008765628.1 | XP_008763850.1 | 3 - 4 | 1859 | 95 | ||
XM_216954.8 | XP_216954.4 | 3 - 4 | 469 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |