Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif3e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011990.1 | NP_001011990.1 | 5 - 6 | 476 | 127 | |
XM_006250385.2 | XP_006250447.1 | 5 - 6 | 518 | 127 | ||
GenBank mRNA | BC082087.1 | - | 5 - 6 | 476 | 7133833 | 127 |
FQ215133.1 | - | 3 - 4 | 287 | 127 | ||
FQ228717.1 | - | 4 - 5 | 287 | 127 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |