Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfng | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199110.1 | NP_954541.1 | 4 - 5 | 663 | 106 | |
GenBank mRNA | BC061801.1 | - | 4 - 5 | 663 | 5598335 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |