Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d22a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006242171.2 | XP_006242233.1 | 6 - 7 | 2648 | 131 | |
XM_008765824.1 | XP_008764046.1 | 6 - 7 | 2646 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |